missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylphenylhydrazine hydrochloride, 97%, Thermo Scientific™

Product Code. 11469547
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469547 5 g 5g
11459547 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469547 Supplier Thermo Scientific Alfa Aesar Supplier No. B22053.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54812-56-5
Molecular Formula C7H10Cl2N2
Molecular Weight (g/mol) 193.071
MDL Number MFCD00012936
InChI Key DQDFVQJOXLGGBL-UHFFFAOYSA-N
Synonym 3-chloro-4-methylphenylhydrazine hydrochloride, 3-chloro-p-tolylhydrazine hydrochloride, 3-chloro-4-methylphenyl hydrazine hydrochloride, 3-chloro-4-methylphenyl hydrazine monohydrochloride, hydrazine, 3-chloro-4-methylphenyl-, monohydrochloride, hydrazine, 3-chloro-4-methylphenyl-, hydrochloride, 3-chloro-4-methylphenyl hydrazine hcl, acmc-1alsi, ksc252c4h, 3-chloro-4-methylphenylhydrazine hcl
PubChem CID 3085075
IUPAC Name (3-chloro-4-methylphenyl)hydrazine;hydrochloride
SMILES CC1=C(C=C(C=C1)NN)Cl.Cl

Specifications

Melting Point ∼233°C (decomposition)
Quantity 5 g
Formula Weight 193.08
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-methylphenylhydrazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.