accessibility menu, dialog, popup

UserName

CAS RN 54812-56-5

IUPAC Name: hydrogen (3-chloro-4-methylphenyl)hydrazine chloride
Synonyms: hydrogen (3-chloro-4-methylphenyl)hydrazine chloride
Molecular Weight (g/mol): 193.07
Molecular Formula: C7H10Cl2N2
InChi Key: DQDFVQJOXLGGBL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=C(NN)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results