Learn More
3-Chloro-2,6-dimethylaniline hydrochloride, 99%, Thermo Scientific™
Product Code. 11414907
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414907 25 g 25g
11404907 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11414907 Supplier Thermo Scientific Alfa Aesar Supplier No. B21851.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 72725-98-5
Molecular Formula C8H11Cl2N
Molecular Weight (g/mol) 192.083
MDL Number MFCD02683075
InChI Key WNBACZJGRRMILU-UHFFFAOYSA-N
Synonym 3-chloro-2,6-dimethylaniline hydrochloride, 3-chloro-2,6-dimethylaniline hcl, 3-chloro-2,6-xylidine hydrochloride, 3-chloro-2,6-dimethyl aniline hydrochloride, 3-chloro-2,6-dimethylaniline-hydrogen chloride 1/1
PubChem CID 12448951
IUPAC Name 3-chloro-2,6-dimethylaniline;hydrochloride
SMILES CC1=C(C(=C(C=C1)Cl)C)N.Cl
Specifications
Melting Point ∼240°C (decomposition)
Quantity 25 g
Formula Weight 192.09
Percent Purity 99%
Chemical Name or Material 3-Chloro-2,6-dimethylaniline hydrochloride