accessibility menu, dialog, popup

UserName

CAS RN 72725-98-5

CH3ClCH3NH2HCl

CAS RN 72725-98-5

IUPAC Name: 3-chloro-2,6-dimethylaniline hydrochloride
Synonyms: 3-chloro-2,6-dimethylaniline hydrochloride
Molecular Weight (g/mol): 192.08
Molecular Formula: C8H11Cl2N
InChi Key: WNBACZJGRRMILU-UHFFFAOYSA-N
SMILES: Cl.CC1=C(N)C(C)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results