missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride, 98%, Thermo Scientific™

Product Code. 11398986
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398986 25 g 25g
11378986 1 g 1g
11388986 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398986 Supplier Thermo Scientific Alfa Aesar Supplier No. L20166.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 292621-58-0
Molecular Formula C7Cl2F4O
Molecular Weight (g/mol) 246.97
MDL Number MFCD03001175
InChI Key GNXAQPALNSLOBK-UHFFFAOYSA-N
Synonym acmc-1cptj, 3-chloro-2,4,5,6-tetrafluorobenzoylchloride, benzoyl chloride,3-chloro-2,4,5,6-tetrafluoro, 3-chloranyl-2,4,5,6-tetrakis fluoranyl benzoyl chloride
PubChem CID 2783249
IUPAC Name 3-chloro-2,4,5,6-tetrafluorobenzoyl chloride
SMILES FC1=C(F)C(Cl)=C(F)C(C(Cl)=O)=C1F

Specifications

Refractive Index 1.4898
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 246.99
Percent Purity 98%
Chemical Name or Material 3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.