CAS RN 292621-58-0
CAS RN 292621-58-0
IUPAC Name:
3-chloro-2,4,5,6-tetrafluorobenzoyl chloride
Synonyms:
3-chloro-2,4,5,6-tetrafluorobenzoyl chloride
Molecular Weight (g/mol):
246.97
Molecular Formula:
C7Cl2F4O
InChi Key:
GNXAQPALNSLOBK-UHFFFAOYSA-N
SMILES:
FC1=C(F)C(Cl)=C(F)C(C(Cl)=O)=C1F
1
–
1
of
1
results
1
3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride, 98%, Thermo Scientific™
CAS: 292621-58-0 Molecular Formula: C7Cl2F4O Molecular Weight (g/mol): 246.97 MDL Number: MFCD03001175 InChI Key: GNXAQPALNSLOBK-UHFFFAOYSA-N Synonym: acmc-1cptj,3-chloro-2,4,5,6-tetrafluorobenzoylchloride,benzoyl chloride,3-chloro-2,4,5,6-tetrafluoro,3-chloranyl-2,4,5,6-tetrakis fluoranyl benzoyl chloride PubChem CID: 2783249 IUPAC Name: 3-chloro-2,4,5,6-tetrafluorobenzoyl chloride SMILES: FC1=C(F)C(Cl)=C(F)C(C(Cl)=O)=C1F