Learn More
3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride, 98%, Thermo Scientific™
Product Code. 11378986
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378986 1 g 1g
11388986 5 g 5g
11398986 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11378986 Supplier Thermo Scientific Alfa Aesar Supplier No. L20166.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 292621-58-0
Molecular Formula C7Cl2F4O
Molecular Weight (g/mol) 246.97
MDL Number MFCD03001175
InChI Key GNXAQPALNSLOBK-UHFFFAOYSA-N
Synonym acmc-1cptj, 3-chloro-2,4,5,6-tetrafluorobenzoylchloride, benzoyl chloride,3-chloro-2,4,5,6-tetrafluoro, 3-chloranyl-2,4,5,6-tetrakis fluoranyl benzoyl chloride
PubChem CID 2783249
IUPAC Name 3-chloro-2,4,5,6-tetrafluorobenzoyl chloride
SMILES FC1=C(F)C(Cl)=C(F)C(C(Cl)=O)=C1F
Specifications
Refractive Index 1.4898
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 246.99
Percent Purity 98%
Chemical Name or Material 3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride