missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothioanisole, 98%, Thermo Scientific Chemicals

Catalog Number 11457086
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
33733-73-2
C7H7BrS
203.097
MFCD00041395
NKYFJZAKUPSUSH-UHFFFAOYSA-N
3-bromothioanisole, 1-bromo-3-methylthio benzene, 3-bromophenyl methyl sulfide, 3-bromophenyl methyl sulfane, 1-bromo-3-methylsulfanyl benzene, benzene, 1-bromo-3-methylthio, m-bromo methylthio benzene, 3-bromophenyl methyl sulphide, 1-bromo-3-methylsulfanyl-benzene, 3-bromothioanisol
2735627
1-bromo-3-methylsulfanylbenzene
CSC1=CC(=CC=C1)Br
This item is not returnable. View return policy
33733-73-2
C7H7BrS
203.097
MFCD00041395
NKYFJZAKUPSUSH-UHFFFAOYSA-N
3-bromothioanisole, 1-bromo-3-methylthio benzene, 3-bromophenyl methyl sulfide, 3-bromophenyl methyl sulfane, 1-bromo-3-methylsulfanyl benzene, benzene, 1-bromo-3-methylthio, m-bromo methylthio benzene, 3-bromophenyl methyl sulphide, 1-bromo-3-methylsulfanyl-benzene, 3-bromothioanisol
2735627
1-bromo-3-methylsulfanylbenzene
CSC1=CC(=CC=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.51
Boiling Point 124°C to 125°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.624
Quantity 1 g
Beilstein 2078598
Formula Weight 203.11
Percent Purity 98%
Chemical Name or Material 3-Bromothioanisole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'