Learn More
3-Bromo-5-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™
Product Code. 15454947
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454947 5 g 5g
15444947 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15454947 Supplier Thermo Scientific Alfa Aesar Supplier No. H32248.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1031929-10-8
Molecular Formula C8H5Br2F3O
Molecular Weight (g/mol) 333.93
MDL Number MFCD11035864
InChI Key QYRNFJCHQHPVTI-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethoxy benzyl bromide, 1-bromo-3-bromomethyl-5-trifluoromethoxy benzene
PubChem CID 26985713
IUPAC Name 1-bromo-3-(bromomethyl)-5-(trifluoromethoxy)benzene
SMILES C1=C(C=C(C=C1OC(F)(F)F)Br)CBr
Specifications
Refractive Index 1.5185
Quantity 5 g
UN Number UN3265
Formula Weight 333.93
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzyl bromide