accessibility menu, dialog, popup

UserName

OFFFBrBr

CAS RN 1031929-10-8

IUPAC Name: 1-bromo-3-(bromomethyl)-5-(trifluoromethoxy)benzene
Synonyms: 1-bromo-3-(bromomethyl)-5-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 333.93
Molecular Formula: C8H5Br2F3O
InChi Key: QYRNFJCHQHPVTI-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC(Br)=CC(CBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results