Learn More
3-Benzoylthiophene-2-carboxylic acid, 96%, Thermo Scientific™
Product Code. 15433817
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
Product Code. Quantity unitSize
15433817 250 mg 250mg
11387569 100 mg 100mg
11307579 1 g 1g
3 options
Product Code. 15433817
Brand: Thermo Scientific Alfa Aesar H28040.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30006-03-2
Molecular Formula C12H8O3S
Molecular Weight (g/mol) 232.25
MDL Number MFCD00159542
InChI Key ZKHLBCVFVQJMBT-UHFFFAOYSA-N
Synonym 3-benzoyl-2-thiophenecarboxylic acid, 3-benzoyl-2-thiophenecarboxylicacid, acmc-1cr5j, 2-thiophenecarboxylicacid, 3-benzoyl, 3-benzoyl-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 3-benzoyl
PubChem CID 315607
IUPAC Name 3-benzoylthiophene-2-carboxylic acid
SMILES OC(=O)C1=C(C=CS1)C(=O)C1=CC=CC=C1
Specifications
Melting Point 150°C to 153°C
Quantity 250 mg
Formula Weight 232.26
Percent Purity 96%
Chemical Name or Material 3-Benzoylthiophene-2-carboxylic acid