accessibility menu, dialog, popup

UserName

CAS RN 30006-03-2

HOOSO

CAS RN 30006-03-2

IUPAC Name: 3-benzoylthiophene-2-carboxylic acid
Synonyms: 3-benzoylthiophene-2-carboxylic acid
Molecular Weight (g/mol): 232.25
Molecular Formula: C12H8O3S
InChi Key: ZKHLBCVFVQJMBT-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CS1)C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results