Learn More
TPP Techno Plastic Products AG 3-B Tissue Culture Test Plate
Click to view available options
No. of Wells:
6
12
24
48
96
Total Volume (Metric) Well:
0.34 mL
0.36 mL
1.49 mL
3.18 mL
5.96 mL
15.53 mL
Unit Size:
Pack of 36
Pack of 50
Description
- The triple bagged packaging (3-B) solution from TPP meets the increasingly more stringent clean room requirements.
- The 3-B products provide noticeably more safety in grade clean aseptic room productions. With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into cleanroom areas.
- The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
- All advantages of the standard product
- Ideal for use in dedicated areas where the sterile transfer of product is essential
- Each product is triple-wrapped in three peel-open bags
- Each bag is marked with a security number: 3 – outer, 2 – middle, 1 – inner bag - the subsequent numbering helps to → show status of packaging
- Innermost bag 1 is labeled with the product data: product number and description, lot number and EXP.
- The safety-open-style peel-open bags are increasing sized, so that during handling of one bag the next inner bag is not accidentally broken rendering it contaminated
Specifications
Specifications
| Packaging | Triple-bagged |
| No. of Wells | 12 |
| Total Volume (Metric) Well | 5.96 mL |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur