Learn More
TPP Techno Plastic Products AG 3-B Tissue Culture Test Plate
Product Code. 17205326
Change view
Click to view available options
No. of Wells:
6
12
24
48
96
Total Volume (Metric) Well:
0.34 mL
0.36 mL
1.49 mL
3.18 mL
5.96 mL
15.53 mL
Unit Size:
Pack of 36
Pack of 50
Product Code. No. of Wells Total Volume (Metric) Well unitSize
17205326 96 0.34 mL Pack of 50
17255316 6 15.53 mL Pack of 36
17265316 12 5.96 mL Pack of 36
17275316 24 3.18 mL Pack of 36
17285316 48 1.49 mL Pack of 36
17295316 96 0.36 mL Pack of 50
6 options
Product Code. 17205326
Brand: TPP Techno Plastic Products AG 392097
Only null left
Add to basket
Add to basket
3-B Tissue culture test plate triple bagged
  • The triple bagged packaging (3-B) solution from TPP meets the increasingly more stringent clean room requirements.
  • The 3-B products provide noticeably more safety in grade clean aseptic room productions. With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into cleanroom areas.
  • The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
  • All advantages of the standard product
  • Ideal for use in dedicated areas where the sterile transfer of product is essential
  • Each product is triple-wrapped in three peel-open bags
  • Each bag is marked with a security number: 3 – outer, 2 – middle, 1 – inner bag - the subsequent numbering helps to → show status of packaging
  • Innermost bag 1 is labeled with the product data: product number and description, lot number and EXP.
  • The safety-open-style peel-open bags are increasing sized, so that during handling of one bag the next inner bag is not accidentally broken rendering it contaminated
Specifications
Packaging Triple-bagged
No. of Wells 96
Total Volume (Metric) Well 0.34 mL