Learn More
TPP Techno Plastic Products AG 3-B Filtermax Flask
3-B Filtermax flask triple bagged
Brand:  TPP Techno Plastic Products AG 399507
Additional Details : Weight : 2.47000kg
 View more versions of this product
Product Code. 17275336
  • 1902.70 NOK / Pack of 12
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • The triple bagged packaging (3-B) solution from TPP meets the increasingly more stringent clean room requirements.
  • The 3-B products provide noticeably more safety in grade clean aseptic room productions. With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into cleanroom areas.
  • The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
  • All advantages of the standard product
  • Ideal for use in dedicated areas where the sterile transfer of product is essential
  • Each product is triple-wrapped in three peel-open bags
  • Each bag is marked with a security number: 3 – outer, 2 – middle, 1 – inner bag - the subsequent numbering helps to show status of packaging
  • Innermost bag 1 is labeled with the product data: product number and description, lot number and EXP.
  • The safety-open-style peel-open bags are increasing sized, so that during handling of one bag the next inner bag is not accidentally broken rendering it contaminated
Specifications
Specifications
Triple-bagged
500 mL