missing translation for 'onlineSavingsMsg'
Learn More

TPP Techno Plastic Products AG 3-B Filtermax Flask

3-B Filtermax flask triple bagged

1902.70 NOK - 9455.20 NOK

Specifications

Packaging Triple-bagged
View More Specs

Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
17245336
View Documents Promotion Details
TPP Techno Plastic Products AG
399157
150 mL
1923.90 NOK
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
 
17255336
View Documents Promotion Details
TPP Techno Plastic Products AG
399257
250 mL
2061.70 NOK
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
 
17275336
View Documents Promotion Details
TPP Techno Plastic Products AG
399507
500 mL
1902.70 NOK
Pack of 12
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
 
17215346
View Documents Promotion Details
TPP Techno Plastic Products AG
399957
1000 mL
9455.20 NOK
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
 
Description

Description

  • The triple bagged packaging (3-B) solution from TPP meets the increasingly more stringent clean room requirements.
  • The 3-B products provide noticeably more safety in grade clean aseptic room productions. With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into cleanroom areas.
  • The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
  • All advantages of the standard product
  • Ideal for use in dedicated areas where the sterile transfer of product is essential
  • Each product is triple-wrapped in three peel-open bags
  • Each bag is marked with a security number: 3 – outer, 2 – middle, 1 – inner bag - the subsequent numbering helps to show status of packaging
  • Innermost bag 1 is labeled with the product data: product number and description, lot number and EXP.
  • The safety-open-style peel-open bags are increasing sized, so that during handling of one bag the next inner bag is not accidentally broken rendering it contaminated
Specifications

Specifications

Triple-bagged
Videos
SDS
Documents

Documents

Product Certifications