missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyridine-4-carboxaldehyde, 95%, Thermo Scientific Chemicals

Product Code. p-7033021
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11474390 250 mg 250mg
11484390 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11474390

Brand: Thermo Scientific Alfa Aesar H50024.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3-Aminopyridine-4-carboxaldehyde is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopyridine-4-carboxaldehyde is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55279-29-3
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD06410684
InChI Key NDEGFXFYOKVWAK-UHFFFAOYSA-N
Synonym 3-aminoisonicotinaldehyde, 3-aminopyridine-4-carboxaldehyde, 3-amino-pyridine-4-carbaldehyde, 3-amino-4-pyridinecarboxaldehyde, 3-amino-4-formylpyridine, 3-amino-4-carboxaldehyde, 4-pyridinecarboxaldehyde, 3-amino, acmc-1b1xn, 3-amino-4-pyridine carboxaldehyde, 4-pyridinecarboxaldehyde,3-amino
PubChem CID 2763001
IUPAC Name 3-aminopyridine-4-carbaldehyde
SMILES NC1=CN=CC=C1C=O

Specifications

Melting Point ∼92°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 122.13
Percent Purity 95%
Chemical Name or Material 3-Aminopyridine-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.