CAS RN 55279-29-3
CAS RN 55279-29-3
IUPAC Name:
3-aminopyridine-4-carbaldehyde
Synonyms:
3-aminopyridine-4-carbaldehyde
Molecular Weight (g/mol):
122.13
Molecular Formula:
C6H6N2O
InChi Key:
NDEGFXFYOKVWAK-UHFFFAOYSA-N
SMILES:
NC1=CN=CC=C1C=O
1
–
1
of
1
results
1
3-Aminopyridine-4-carboxaldehyde, 95%, Thermo Scientific Chemicals
CAS: 55279-29-3 Molecular Formula: C6H6N2O Molecular Weight (g/mol): 122.13 MDL Number: MFCD06410684 InChI Key: NDEGFXFYOKVWAK-UHFFFAOYSA-N Synonym: 3-aminoisonicotinaldehyde,3-aminopyridine-4-carboxaldehyde,3-amino-pyridine-4-carbaldehyde,3-amino-4-pyridinecarboxaldehyde,3-amino-4-formylpyridine,3-amino-4-carboxaldehyde,4-pyridinecarboxaldehyde, 3-amino,acmc-1b1xn,3-amino-4-pyridine carboxaldehyde,4-pyridinecarboxaldehyde,3-amino PubChem CID: 2763001 IUPAC Name: 3-aminopyridine-4-carbaldehyde SMILES: NC1=CN=CC=C1C=O