Learn More
3-Allylsalicylaldehyde, 97%
Product Code. 15480328
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
15480328 5 g 5g
15490328 25 g 25g
2 options
Product Code. 15480328
Brand: Thermo Scientific Alfa Aesar H53475.06
Request Bulk or Custom Format
CAS: 24019-66-7 | C10H10O2 | 162.188 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
3-Allylsalicylaldehyde is a synthetic intermediate for the synthesis of 3-aminoflavones. Flavonoids have been studied for their antiproliferative activity and in vitro cytotoxicity. It is used in the preparation of oxazole derivatives and also imidazole scaffold-based 2-substituted benzofurans for application against human tumor cell lines. It is a reagent in the synthesis of antioxidant and antimicrobial novel chalcones.
Solubility
Soluble in chloroform.
Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24019-66-7
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00037364
InChI Key INLWEXRRMUMHKB-UHFFFAOYSA-N
Synonym 3-allyl-2-hydroxybenzaldehyde, 3-allylsalicylaldehyde, 3-allyl-2-hydroxy-benzaldehyde, benzaldehyde, 2-hydroxy-3-2-propenyl, salicylaldehyde, 3-allyl, 2-hydroxy-3-prop-2-en-1-yl benzaldehyde, acmc-209wev, tpc-sk001, 2-hydroxy-3-allylbenzaldehyde
PubChem CID 141062
IUPAC Name 2-hydroxy-3-prop-2-enylbenzaldehyde
SMILES C=CCC1=CC=CC(=C1O)C=O
Specifications
Density 1.099
Flash Point 107°C (225°F)
Refractive Index 1.5645
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform.
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 3-Allylsalicylaldehyde