accessibility menu, dialog, popup

UserName

CAS RN 24019-66-7

OHCH2O

CAS RN 24019-66-7

IUPAC Name: 2-hydroxy-3-(prop-2-en-1-yl)benzaldehyde
Synonyms: 2-hydroxy-3-(prop-2-en-1-yl)benzaldehyde
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: INLWEXRRMUMHKB-UHFFFAOYSA-N
SMILES: OC1=C(CC=C)C=CC=C1C=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)