missing translation for 'onlineSavingsMsg'
Learn More

3-(Acetamidomethyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15490008
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15490008 1 g 1g
15400018 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15490008

Brand: Thermo Scientific Alfa Aesar H53252.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(Acetamidomethyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-42-2
Molecular Formula C9H12BNO3
Molecular Weight (g/mol) 193.009
MDL Number MFCD06659819
InChI Key IXRGNMIMGAJHEG-UHFFFAOYSA-N
Synonym 3-acetamidomethyl phenyl boronic acid, 3-acetamidomethylphenylboronic acid, 3-acetylaminomethyl benzeneboronic acid, 3-acetamidomethyl phenylboronic acid, 3-acetamidomethyl benzeneboronic acid, boronic acid,b-3-acetylamino methyl phenyl, pubchem14581, acmc-209q1y
PubChem CID 22309455
IUPAC Name [3-(acetamidomethyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)CNC(=O)C)(O)O

Specifications

Melting Point 253°C to 255°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 3-(Acetamidomethyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.