accessibility menu, dialog, popup

UserName

CAS RN 850568-42-2

BHNOCH3HOOH

CAS RN 850568-42-2

IUPAC Name: [3-(acetamidomethyl)phenyl]boronic acid
Synonyms: 3-(acetamidomethyl)phenylboronic acid
Molecular Weight (g/mol): 193.01
Molecular Formula: C9H12BNO3
InChi Key: IXRGNMIMGAJHEG-UHFFFAOYSA-N
SMILES: CC(=O)NCC1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results