Learn More
3,5-Dichlorotoluene, TRC
Product Code. 30009275
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009275 5 g 5g
30011580 2.5 g 2.5g
17983405 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009275 Supplier Toronto Research Chemicals Supplier No. TRCD4368355G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25186-47-4
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.03
Synonym Toluene, 3,5-dichloro- (8CI), 1,3-Dichloro-5-methylbenzene, 3,5-Dichlorotoluene, 1,3-Dichloro-5-methylbenzene, Toluene, 3,5-dichloro- (8CI)
IUPAC Name 1,3-dichloro-5-methylbenzene
SMILES Cc1cc(Cl)cc(Cl)c1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C7H6Cl2/c1-5-2-6(8)4-7(9)3-5/h2-4H,1H3
Formula Weight 159.9847
Chemical Name or Material 3,5-Dichlorotoluene