Learn More
3-[4-Fluoro-3-(trifluoromethoxy)phenyl]propionic acid, 97%, Thermo Scientific™
Product Code. 15498745
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15498745
Brand: Thermo Scientific Alfa Aesar H34036.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1240256-80-7
Molecular Formula C10H8F4O3
Molecular Weight (g/mol) 252.165
MDL Number MFCD09999449
InChI Key RDAQINCBAFDESL-UHFFFAOYSA-N
Synonym 3-4-fluoro-3-trifluoromethoxy phenyl propionic acid, 3-4-fluoro-3-trifluoromethoxy phenyl propanoic acid, 3-4-fluoro-3-trifluoromethoxy phenyl propanoic a
PubChem CID 66523471
IUPAC Name 3-[4-fluoro-3-(trifluoromethoxy)phenyl]propanoic acid
SMILES C1=CC(=C(C=C1CCC(=O)O)OC(F)(F)F)F
Specifications
Quantity 1 g
Formula Weight 252.16
Percent Purity 97%
Chemical Name or Material 3-[4-Fluoro-3-(trifluoromethoxy)phenyl]propionic acid