CAS RN 1240256-80-7
CAS RN 1240256-80-7
IUPAC Name:
3-[4-fluoro-3-(trifluoromethoxy)phenyl]propanoic acid
Synonyms:
3-[4-fluoro-3-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol):
252.17
Molecular Formula:
C10H8F4O3
InChi Key:
RDAQINCBAFDESL-UHFFFAOYSA-N
SMILES:
OC(=O)CCC1=CC(OC(F)(F)F)=C(F)C=C1
1
–
1
of
1
results
1
3-[4-Fluoro-3-(trifluoromethoxy)phenyl]propionic acid, 97%, Thermo Scientific™
CAS: 1240256-80-7 Molecular Formula: C10H8F4O3 Molecular Weight (g/mol): 252.165 MDL Number: MFCD09999449 InChI Key: RDAQINCBAFDESL-UHFFFAOYSA-N Synonym: 3-4-fluoro-3-trifluoromethoxy phenyl propionic acid,3-4-fluoro-3-trifluoromethoxy phenyl propanoic acid,3-4-fluoro-3-trifluoromethoxy phenyl propanoic a PubChem CID: 66523471 IUPAC Name: 3-[4-fluoro-3-(trifluoromethoxy)phenyl]propanoic acid SMILES: C1=CC(=C(C=C1CCC(=O)O)OC(F)(F)F)F