Learn More
3,4-Dimethylbenzotrifluoride, TRC
Product Code. 30377589
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377589 100 mg -
30369028 50 mg -
30371500 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30377589 Supplier Toronto Research Chemicals Supplier No. TRCD462098100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78164-31-5
Molecular Formula C9H9F3
Molecular Weight (g/mol) 174.16
SMILES Cc1ccc(cc1C)C(F)(F)F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C9H9F3/c1-6-3-4-8(5-7(6)2)9(10,11)12/h3-5H,1-2H3
Formula Weight 174.0656
Chemical Name or Material 3,4-Dimethylbenzotrifluoride