CAS RN 78164-31-5
IUPAC Name:
1,2-dimethyl-4-(trifluoromethyl)benzene
Synonyms:
1,2-dimethyl-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
174.17
Molecular Formula:
C9H9F3
InChi Key:
XBHTYKIYEUHMIA-UHFFFAOYSA-N
SMILES:
CC1=C(C)C=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
3,4-Dimethylbenzotrifluoride, TRC
CAS: 78164-31-5 Molecular Formula: C9H9F3 Molecular Weight (g/mol): 174.16 SMILES: Cc1ccc(cc1C)C(F)(F)F