missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-L-phenylalanine, 98%, Thermo Scientific™

Product Code. 15493707
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493707 250 mg 250mg
11411830 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493707 Supplier Thermo Scientific Alfa Aesar Supplier No. H27539.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31105-90-5
Molecular Formula C9H9F2NO2
Molecular Weight (g/mol) 201.17
MDL Number MFCD01075125
InChI Key PRAWYXDDKCVZTL-QMMMGPOBSA-N
Synonym 3,4-difluoro-l-phenylalanine, s-2-amino-3-3,4-difluorophenyl propanoic acid, l-3,4-difluorophenylalanine, 2s-2-amino-3-3,4-difluorophenyl propanoic acid, l-3,4-difluorophe, phenylalanine, 3,4-difluoro, h-phe 3,4-f2-oh, pubchem17977, h-phe 3,4-dif-oh, l-phe 3,4-dif-oh
PubChem CID 716295
IUPAC Name (2S)-2-amino-3-(3,4-difluorophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(F)C(F)=C1)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 201.18
Percent Purity 98%
Chemical Name or Material 3,4-Difluoro-L-phenylalanine
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.