accessibility menu, dialog, popup

UserName

CAS RN 31105-90-5

NH2(S)FFOHO

CAS RN 31105-90-5

IUPAC Name: (2S)-2-amino-3-(3,4-difluorophenyl)propanoic acid
Synonyms: (2S)-2-amino-3-(3,4-difluorophenyl)propanoic acid
Molecular Weight (g/mol): 201.17
Molecular Formula: C9H9F2NO2
InChi Key: PRAWYXDDKCVZTL-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=C(F)C(F)=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)