Learn More
3-(4-Biphenylyl)-N-Fmoc-D-alanine, 95%
Product Code. 15488627
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488627 5 g 5g
15498627 250 mg 250mg
15478627 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15488627 Supplier Thermo Scientific Alfa Aesar Supplier No. H52108.06
Request Bulk or Custom Format
CAS: 205526-38-1 | C30H25NO4 | 463.53 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 205526-38-1
Molecular Formula C30H25NO4
Molecular Weight (g/mol) 463.53
MDL Number MFCD00191199
InChI Key VSGACONKQRJFGX-UHFFFAOYNA-N
Synonym fmoc-d-4,4'-biphenylalanine, fmoc-d-bip-oh, fmoc-d-bip 4,4'-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1,1'-biphenyl-4-yl propanoic acid, fmoc-4-biphenyl-d-ala, fmoc-l-phe 4-ph-oh, fmoc-bip-oh, 2r-3-1,1'-biphenyl-4-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1664, fmoc-d-bip 4,4atrade mark-oh
PubChem CID 7019142
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylphenyl)propanoic acid
SMILES OC(=O)C(CC1=CC=C(C=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Specifications
Quantity 5 g
Formula Weight 463.52
Percent Purity 95%
Chemical Name or Material 3-(4-Biphenylyl)-N-Fmoc-D-alanine