CAS RN 205526-38-1
CAS RN 205526-38-1
IUPAC Name:
3-{[1,1'-biphenyl]-4-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms:
3-{[1,1'-biphenyl]-4-yl}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol):
463.53
Molecular Formula:
C30H25NO4
InChi Key:
VSGACONKQRJFGX-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CC=C(C=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
3-(4-Biphenylyl)-N-Fmoc-D-alanine, 95%
CAS: 205526-38-1 Molecular Formula: C30H25NO4 Molecular Weight (g/mol): 463.53 MDL Number: MFCD00191199 InChI Key: VSGACONKQRJFGX-UHFFFAOYNA-N Synonym: fmoc-d-4,4'-biphenylalanine,fmoc-d-bip-oh,fmoc-d-bip 4,4'-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1,1'-biphenyl-4-yl propanoic acid,fmoc-4-biphenyl-d-ala,fmoc-l-phe 4-ph-oh,fmoc-bip-oh,2r-3-1,1'-biphenyl-4-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,ambotzfaa1664,fmoc-d-bip 4,4atrade mark-oh PubChem CID: 7019142 IUPAC Name: (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylphenyl)propanoic acid SMILES: OC(=O)C(CC1=CC=C(C=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12