missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trichloroaniline, 97%, Thermo Scientific™

Product Code. 11471427
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11471427

Brand: Thermo Scientific Alfa Aesar B24927.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,4,5-Trichloroaniline was used in the determination of substituted aromatic amines in sediment or water samples by capillary column gas chromatography.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-91-3
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.46
MDL Number MFCD00007769
InChI Key XOGYQVITULCUGU-UHFFFAOYSA-N
Synonym benzenamine, 3,4,5-trichloro, ccris 2398, aniline, 3,4,5-trichloro, 3,4,5 trichloroaniline, acmc-1bag0, 3,4,5-trichloro-aniline, 3,4,5-trichlorophenylamine, 3,4,5-trichlorobenzenamine, 3,4,5-tris chloranyl aniline, 3,4,5-trichloroaniline
PubChem CID 12469
IUPAC Name 3,4,5-trichloroaniline
SMILES NC1=CC(Cl)=C(Cl)C(Cl)=C1

Specifications

Melting Point 98°C to 100°C
Quantity 25 g
UN Number UN2018
Solubility Information Insoluble in water.
Formula Weight 196.46
Percent Purity 97%
Chemical Name or Material 3,4,5-Trichloroaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.