accessibility menu, dialog, popup

UserName

CAS RN 634-91-3

NH2ClClCl

CAS RN 634-91-3

IUPAC Name: 3,4,5-trichloroaniline
Synonyms: 3,4,5-trichloroaniline
Molecular Weight (g/mol): 196.46
Molecular Formula: C6H4Cl3N
InChi Key: XOGYQVITULCUGU-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(Cl)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results