missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthylthio)propionic acid, 97%, Thermo Scientific™

Catalog Number 11342416
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1141-45-3
C13H12O2S
232.30
MFCD00051616
XMQJAJQNGNSZFY-UHFFFAOYSA-N
3-2-naphthylthio propionic acid, 3-naphthalen-2-ylthio propanoic acid, 3-2-naphthylthio propanoic acid, 3-2-naphthylsulfanyl propanoic acid, 3-naphthalen-2-ylsulfanyl propanoic acid, maybridge3_005839, acmc-1ca98, 3-2-naphthylsulfanyl propanoic acid #, 3-naphthalen-2-ylsulfanyl-propionic acid
517992
3-naphthalen-2-ylsulfanylpropanoic acid
OC(=O)CCSC1=CC2=CC=CC=C2C=C1
This item is not returnable. View return policy
1141-45-3
C13H12O2S
232.30
MFCD00051616
XMQJAJQNGNSZFY-UHFFFAOYSA-N
3-2-naphthylthio propionic acid, 3-naphthalen-2-ylthio propanoic acid, 3-2-naphthylthio propanoic acid, 3-2-naphthylsulfanyl propanoic acid, 3-naphthalen-2-ylsulfanyl propanoic acid, maybridge3_005839, acmc-1ca98, 3-2-naphthylsulfanyl propanoic acid #, 3-naphthalen-2-ylsulfanyl-propionic acid
517992
3-naphthalen-2-ylsulfanylpropanoic acid
OC(=O)CCSC1=CC2=CC=CC=C2C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 103°C to 107°C
Quantity 1 g
Beilstein 2558498
Formula Weight 232.3
Percent Purity 97%
Chemical Name or Material 3-(2-Naphthylthio)propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'