missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus VP16 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-950-0.025ml
This item is not returnable.
View return policy
Description
HSV VP16 Polyclonal specifically detects HSV VP16 in Virus samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| HSV VP16 | |
| Polyclonal | |
| Unconjugated | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| Synthetic peptide corresponding to amino acids 474-487 of the herpes simplex virus HSV VP16 protein, conjugated to KLH via an N-terminal added cysteine. | |
| 0.025 mL | |
| Primary | |
| Virus | |
| IgG |
| Western Blot | |
| 5.8 mg/mL | |
| Western Blot 1:1000, Immunoprecipitation 10ug | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Recognizes HSV VP16 fusion protein. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction