missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Herpes Simplex Virus VP16 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
1985.00 NOK
Specifications
| Antigen | HSV VP16 |
|---|---|
| Concentration | 5.8 mg/mL |
| Dilution | Western Blot 1:1000, Immunoprecipitation 10ug |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
HSV VP16 Polyclonal specifically detects HSV VP16 in Virus samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| HSV VP16 | |
| Western Blot 1:1000, Immunoprecipitation 10ug | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| Synthetic peptide corresponding to amino acids 474-487 of the herpes simplex virus HSV VP16 protein, conjugated to KLH via an N-terminal added cysteine. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 5.8 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| IgG | |
| Affinity Purified | |
| Recognizes HSV VP16 fusion protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title