Learn More
Endecotts™ Stainless Steel Test Sieve, 8 in. dia.
Product Code. 10759235
Change view
Click to view available options
Pore Size:
106 μm
250 μm
425 μm
500 μm
Unit Size:
Each
Pack of 16
Product Code. Pore Size unitSize
10759235 425 μm Each
10758855 106 μm Each
11532005 106 μm Pack of 16
10441734 500 μm Each
11541995 500 μm Pack of 16
11551995 425 μm Pack of 16
11581995 250 μm Pack of 16
11326030 250 μm Each
8 options
Product Code. 10759235
Brand: Endecotts™ 008SAW.425
Only null left
Add to basket
Add to basket
Full height stainless steel test sieves offer all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Test Sieve, 8 in. dia. complies with ASTM 11 standards
  • Stainless steel precision frame (ensures consistent nestability)
  • Stainless steel mesh
  • Precise aperture in accordance with ISO 3310-1 / BS410
  • Natural fillet (free flowing of sample)
  • Totally sealed (no crevice to lose material)
  • Evenly tensioned mesh (ensures accurate analysis)
  • Safe edge (big radius makes it comfortable to handle)
  • Serial number (ensures full traceability)
Specifications
Quantity 1Each
Pore Size 425 μm
Type Test Sieve
Certifications/Compliance ISO 3310-1 & 2 requirements, ASTM, BSI
Material Stainless Steel
For Use With (Application) For all types of laboratory sampling and particle size analysis
Diameter (Metric) Outer 203 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel
Sieve Number 40