Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ISO Certificate, Pore Sizes: Under 10 mm
Product Code. 11974482 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.12 mm
1.18 mm
1.25 mm
1.40 mm
1.60 mm
1.70 mm
1.80 mm
2 mm
2.24 mm
2.36 mm
2.50 mm
2.80 mm
3.15 mm
3.35 mm
3.55 mm
4 mm
4.50 mm
4.75 mm
5 mm
5.60 mm
6.30 mm
6.70 mm
7.10 mm
8 mm
9 mm
9.50 mm
Unit Size:
Each
Product Code. Pore Size unitSize
11974482 1.12 mm Each
11904492 1.18 mm Each
11984482 1.40 mm Each
11994482 1.25 mm Each
11914492 1.60 mm Each
11924492 1.70 mm Each
11964482 1 mm Each
11934492 1.80 mm Each
11954492 2.24 mm Each
11944492 2 mm Each
11964492 2.36 mm Each
11984492 2.80 mm Each
11974492 2.50 mm Each
11994492 3.15 mm Each
11904502 3.35 mm Each
11934502 4.75 mm Each
11944502 4.50 mm Each
11914502 3.55 mm Each
11924502 4 mm Each
11974502 5.60 mm Each
11954502 5 mm Each
11984502 6.70 mm Each
11994502 7.10 mm Each
11964502 6.30 mm Each
11904512 8 mm Each
11924512 9.50 mm Each
11914512 9 mm Each
27 options
Product Code. 11974482
Brand: RETSCH 60.140.001120
Only null left
Specifications
Quantity 1Each
Pore Size 1.12 mm
Type Test Sieve
Certifications/Compliance ISO 3310/1
Material Stainless Steel
For Use With (Equipment) Shaker AS 300
Height (Metric) 25.4mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 203.2 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel