Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ASTM Certificate, Micrometer Pore Sizes
Product Code. 11954632 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
106 μm
125 μm
150 μm
180 μm
212 μm
250 μm
300 μm
355 μm
425 μm
500 μm
600 μm
710 μm
850 μm
Unit Size:
Each
Product Code. Pore Size unitSize
11954632 25 μm Each
11934642 106 μm Each
11944642 125 μm Each
11944632 20 μm Each
11974642 212 μm Each
11954642 180 μm Each
11964642 150 μm Each
11984642 250 μm Each
11994642 300 μm Each
11904652 355 μm Each
11964632 32 μm Each
11974632 38 μm Each
11984632 45 μm Each
11934652 500 μm Each
11914652 425 μm Each
11994632 53 μm Each
11944652 600 μm Each
11954652 710 μm Each
11904642 75 μm Each
11914642 63 μm Each
11924642 90 μm Each
11964652 850 μm Each
22 options
Product Code. 11954632
Brand: RETSCH 60.141.000025
Only null left
Specifications
Quantity 1Each
Pore Size 25 μm
Type Test Sieve
Certifications/Compliance ASTM E 11
Material Stainless Steel
For Use With (Equipment) Shaker AS 300
Height (Metric) 25.4mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 203.2 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel