Learn More
Eppendorf™ 2000μL Sterile Polypropylene 96-Well Deep Well PCR Plates
BIOPHARMAGREENER_CHOICE
Product Code. 10227512 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Green
White
Yellow
Packaging:
10 Bags x 8 Plates
5 Bags x 4 Plates
Unit Size:
Pack of 20
Pack of 80
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Packaging unitSize
10227512 Blue 5 Bags x 4 Plates Pack of 20
10165412 Green 5 Bags x 4 Plates Pack of 20
10359192 Yellow 5 Bags x 4 Plates Pack of 20
10052143 White 10 Bags x 8 Plates Pack of 80
10267462 White 5 Bags x 4 Plates Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10227512 Supplier Eppendorf™ Supplier No. 0030502345
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
PCR plates, free of DNA, DNase, RNase and PCR inhibitors
These plates combine a new well design with a sophisticated color coding system for ease of reading and orientation. The well geometry allows high speed pipetting without splashing, excellent mixing and high speed centrifugation, which accelerates your manual and automated work processes. The new polypropylene plates fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines.
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • Free from DNA, DNase, RNase and PCR* inhibitor (PCR* clean)
  • Autoclavable
  • Optionally available with barcode
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Packaging 5 Bags x 4 Plates
Color Blue
Material Polypropylene
No. of Wells 96