Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Product Code. 10780853 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.18 mm
1.4 mm
1.7 mm
2 mm
2.36 mm
2.8 mm
3.35 mm
355 μm
4 mm
425 μm
4.75 mm
500 μm
5.6 mm
600 μm
6.3 mm
6.7 mm
710 μm
8 mm
850 μm
9.5 mm
11.2 mm
12.5 mm
13.2 mm
16 mm
19 mm
22.4 mm
25 mm
26.5 mm
31.5 mm
37.5 mm
45 mm
50 mm
53 mm
63 mm
75 mm
90 mm
100 mm
106 mm
125 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10780853 26.5 mm Each
10574682 1.18 mm Each
10196261 1.4 mm Each
10106271 1.7 mm Each
10238781 125 mm Each
10075001 12.5 mm Each
10417771 100 mm Each
10390662 106 mm Each
10660282 11.2 mm Each
10186831 13.2 mm Each
10126931 16 mm Each
10035041 19 mm Each
10660272 1 mm Each
10300292 2.36 mm Each
10515072 2.8 mm Each
10268641 22.4 mm Each
10446431 25 mm Each
10116271 2 mm Each
10054921 3.35 mm Each
10790853 31.5 mm Each
10709132 4.75 mm Each
10196021 355 μm Each
10600102 425 μm Each
10544112 37.5 mm Each
10127221 45 mm Each
10610282 4 mm Each
10300342 5.6 mm Each
10300722 500 μm Each
10427201 50 mm Each
10498151 6.7 mm Each
10004961 6.3 mm Each
10340862 600 μm Each
10594682 63 mm Each
10320772 53 mm Each
10005041 850 μm Each
10650282 8 mm Each
10638571 75 mm Each
10678751 90 mm Each
10526222 9.5 mm Each
10650272 710 μm Each
40 options
Product Code. 10780853
Brand: RETSCH 60.123.026500
Only null left
Specifications
Quantity 1Each
Pore Size 26.5 mm
Type Test Sieve
Certifications/Compliance ASTM E 11
Material Stainless Steel
For Use With (Equipment) Shaker AS 300
Height (Metric) 25 mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel