Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Product Code. 10638571 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.18 mm
1.4 mm
1.7 mm
2 mm
2.36 mm
2.8 mm
3.35 mm
355 μm
4 mm
425 μm
4.75 mm
500 μm
5.6 mm
600 μm
6.3 mm
6.7 mm
710 μm
8 mm
850 μm
9.5 mm
11.2 mm
12.5 mm
13.2 mm
16 mm
19 mm
22.4 mm
25 mm
26.5 mm
31.5 mm
37.5 mm
45 mm
50 mm
53 mm
63 mm
75 mm
90 mm
100 mm
106 mm
125 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10638571 | 75 mm | Each |
| 10574682 | 1.18 mm | Each |
| 10196261 | 1.4 mm | Each |
| 10106271 | 1.7 mm | Each |
| 10238781 | 125 mm | Each |
| 10075001 | 12.5 mm | Each |
| 10417771 | 100 mm | Each |
| 10390662 | 106 mm | Each |
| 10660282 | 11.2 mm | Each |
| 10186831 | 13.2 mm | Each |
| 10126931 | 16 mm | Each |
| 10035041 | 19 mm | Each |
| 10660272 | 1 mm | Each |
| 10300292 | 2.36 mm | Each |
| 10515072 | 2.8 mm | Each |
| 10268641 | 22.4 mm | Each |
| 10446431 | 25 mm | Each |
| 10116271 | 2 mm | Each |
| 10780853 | 26.5 mm | Each |
| 10054921 | 3.35 mm | Each |
| 10790853 | 31.5 mm | Each |
| 10709132 | 4.75 mm | Each |
| 10196021 | 355 μm | Each |
| 10600102 | 425 μm | Each |
| 10544112 | 37.5 mm | Each |
| 10127221 | 45 mm | Each |
| 10610282 | 4 mm | Each |
| 10300342 | 5.6 mm | Each |
| 10300722 | 500 μm | Each |
| 10427201 | 50 mm | Each |
| 10498151 | 6.7 mm | Each |
| 10004961 | 6.3 mm | Each |
| 10340862 | 600 μm | Each |
| 10594682 | 63 mm | Each |
| 10320772 | 53 mm | Each |
| 10005041 | 850 μm | Each |
| 10650282 | 8 mm | Each |
| 10678751 | 90 mm | Each |
| 10526222 | 9.5 mm | Each |
| 10650272 | 710 μm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 75 mm |
| Type | Test Sieve |
| Certifications/Compliance | ASTM E 11 |
| Material | Stainless Steel |
| For Use With (Equipment) | Shaker AS 300 |
| Height (Metric) | 25 mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 200 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |
| Show More Show Less |