Learn More
Fisherbrand™ Two-Way Scaffolding Connectors
Product Code. 11518112 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Product Type:
2-Way Connector
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11518112 2-Way Connector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11518112
In Stock
Add to basket
Add to basket
Used for fixing two rods or tubes at right angles. Fisherbrand™ Two-Way Scaffolding Connectors can be tightened by hand.
  • With thumbscrews
  • For rods and tubes of 6 to 13mm diameters
Especificaciones
Product Type 2-Way Connector
For Use With (Application) 2-Way
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.