Learn More
2-(N-Ethylanilino)ethanol, TRC
Product Code. 30384607
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384607 25 g
30376771 5 g
30379757 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30384607 Supplier Toronto Research Chemicals Supplier No. TRCE67876525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 92-50-2
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.23
Synonym 2-(Ethylphenylamino)ethanol, Ethylanilinoethanol, Ethylphenylethanolamine, Hydroxyethylethylaniline, N-(2-Hydroxyethyl)-N-ethylaniline, Phenylethylethanolamine
IUPAC Name 2-(N-ethylanilino)ethanol
SMILES CCN(CCO)c1ccccc1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C10H15NO/c1-2-11(8-9-12)10-6-4-3-5-7-10/h3-7,12H,2,8-9H2,1H3
Formula Weight 165.1154
Chemical Name or Material 2-(N-Ethylanilino)ethanol