accessibility menu, dialog, popup

UserName

CAS RN 92-50-2

H3CNOH

CAS RN 92-50-2

IUPAC Name: 2-[ethyl(phenyl)amino]ethan-1-ol
Synonyms: ethanol, 2-(ethylphenylamino)-
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: HYVGFUIWHXLVNV-UHFFFAOYSA-N
SMILES: CCN(CCO)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)