missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-(4-pyridyl)thiazole, 97%, Thermo Scientific™

Product Code. 15448687
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448687 5 g 5g
15438687 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448687 Supplier Thermo Scientific Alfa Aesar Supplier No. H52155.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23899-32-3
Molecular Formula C9H8N2S
Molecular Weight (g/mol) 176.24
MDL Number MFCD16658845
InChI Key SZDNUULQXPIYIT-UHFFFAOYSA-N
Synonym 2-methyl-4-4-pyridyl thiazole, 4-2-methyl-1,3-thiazol-4-yl pyridine, 2-methyl-4-pyridin-4-yl thiazole
PubChem CID 17896320
SMILES CC1=NC(=CS1)C1=CC=NC=C1

Specifications

Quantity 5 g
IUPAC Name 2-methyl-4-pyridin-4-yl-1,3-thiazole
Formula Weight 176.24
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-(4-pyridyl)thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.