accessibility menu, dialog, popup

UserName

CAS RN 23899-32-3

CH3NSN

CAS RN 23899-32-3

IUPAC Name: 4-(2-methyl-1,3-thiazol-4-yl)pyridine
Synonyms: 4-(2-methyl-1,3-thiazol-4-yl)pyridine
Molecular Weight (g/mol): 176.24
Molecular Formula: C9H8N2S
InChi Key: SZDNUULQXPIYIT-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results