Learn More
2-Fluoro-6-(trifluoromethyl)aniline, 98%, Thermo Scientific™
Product Code. 11479147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479147 5 g 5g
11469147 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11479147 Supplier Thermo Scientific Alfa Aesar Supplier No. B20684.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144851-61-6
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.12
MDL Number MFCD00061242
InChI Key CQSFHEFEKDRLKE-UHFFFAOYSA-N
Synonym 2-fluoro-6-trifluoromethyl aniline, 2-amino-3-fluorobenzotrifluoride, benzenamine, 2-fluoro-6-trifluoromethyl, 2-fluoro-6-trifluoromethyl-phenylamine, 2-fluoro-6-trifluoromethyl phenylamine, pubchem2765, acmc-20aid6, intermediates-zcf02111, 2-fluoro-6-trifluoromethyl-aniline
PubChem CID 518959
IUPAC Name 2-fluoro-6-(trifluoromethyl)aniline
SMILES NC1=C(F)C=CC=C1C(F)(F)F
Specifications
Density 1.388
Boiling Point 155°C
Flash Point 53°C (127°F)
Refractive Index 1.461
Quantity 5 g
UN Number UN1992
Formula Weight 179.12
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)aniline