CAS RN 144851-61-6
CAS RN 144851-61-6
IUPAC Name:
2-fluoro-6-(trifluoromethyl)aniline
Synonyms:
2-fluoro-6-(trifluoromethyl)aniline
Molecular Weight (g/mol):
179.12
Molecular Formula:
C7H5F4N
InChi Key:
CQSFHEFEKDRLKE-UHFFFAOYSA-N
SMILES:
NC1=C(F)C=CC=C1C(F)(F)F
1
–
1
of
1
results
1
2-Fluoro-6-(trifluoromethyl)aniline, 98%, Thermo Scientific™
CAS: 144851-61-6 Molecular Formula: C7H5F4N Molecular Weight (g/mol): 179.12 MDL Number: MFCD00061242 InChI Key: CQSFHEFEKDRLKE-UHFFFAOYSA-N Synonym: 2-fluoro-6-trifluoromethyl aniline,2-amino-3-fluorobenzotrifluoride,benzenamine, 2-fluoro-6-trifluoromethyl,2-fluoro-6-trifluoromethyl-phenylamine,2-fluoro-6-trifluoromethyl phenylamine,pubchem2765,acmc-20aid6,intermediates-zcf02111,2-fluoro-6-trifluoromethyl-aniline PubChem CID: 518959 IUPAC Name: 2-fluoro-6-(trifluoromethyl)aniline SMILES: NC1=C(F)C=CC=C1C(F)(F)F