missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15497605
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497605 250 mg 250mg
11372619 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497605 Supplier Thermo Scientific Alfa Aesar Supplier No. H26687.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride is used to describe a straight or branched chain alkyl.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1744-43-0
Molecular Formula C7H3ClF4O2S
Molecular Weight (g/mol) 262.603
MDL Number MFCD01090999
InChI Key XTHDSFNKYHMRFZ-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl benzenesulfonyl chloride, 2-fluoro-5-trifluoromethyl benzene-1-sulfonyl chloride, 2-fluoro-5-trifluoromethyl benzenesulphonyl chloride, 2-fluoro-5-trifluoromethylphenylsulfonyl chloride, pubchem8505, acmc-20anv2, chloro 2-fluoro-5-trifluoromethyl phenyl sulfone, 2-fluoro-5-trifluoromethyl phenylsulfonyl chloride, 2-fluoro-5-trifluoromethyl-benzenesulfonyl chloride, 2-fluoranyl-5-trifluoromethyl benzenesulfonyl chloride
PubChem CID 2737547
IUPAC Name 2-fluoro-5-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1C(F)(F)F)S(=O)(=O)Cl)F

Tekniske data

Refractive Index 1.478
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 262.61
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.